siRNA Details
Gene Symbol: Mta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171052.1 | NP_001164523.1 | 3 | 251 | ||
NM_001171053.1 | NP_001164524.1 | 3 | 251 | |||
NM_001171054.1 | NP_001164525.1 | 3 | 251 | |||
NM_054082.2 | NP_473423.1 | 3 | 251 | |||
GenBank mRNA | AF288138.1 | - | 3 | 205 | ||
AK129318.1 | - | 3 | 180 | |||
AK145942.1 | - | 3 | 196 | |||
AK146721.1 | - | 3 | 204 | |||
AK146905.1 | - | 3 | 204 | |||
AK147197.1 | - | 3 | 194 | |||
AK154856.1 | - | 3 | 191 | |||
AK158866.1 | - | 2 | 114 | |||
BC022124.1 | - | 3 | 215 | 5097283 | ||
BC099960.1 | - | 3 | 168 | 30023887 |
