siRNA Details
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 14 | 1793 | ||
GenBank mRNA | AF244362.3 | - | 13 | 1428 | ||
AK088518.1 | - | 13 | 1489 | |||
BC023956.1 | - | 14 | 1562 | 5321098 | ||
BC034059.1 | - | 8 | 797 | 4163877 | ||
BC096743.1 | - | 14 | 1816 | 30549276 |
