siRNA Details
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 11 | 1381 | ||
GenBank mRNA | AF244362.3 | - | 10 | 1016 | ||
AK088518.1 | - | 10 | 1077 | |||
AK216199.1 | - | 2 | 133 | |||
AK216883.1 | - | 1 | 37 | |||
BC023956.1 | - | 11 | 1150 | 5321098 | ||
BC034059.1 | - | 5 | 385 | 4163877 | ||
BC096743.1 | - | 11 | 1404 | 30549276 |
