siRNA Details
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 16 | 2203 | ||
GenBank mRNA | AF244362.3 | - | 15 | 1838 | ||
AK088518.1 | - | 15 | 1899 | |||
BC023956.1 | - | 16 | 1972 | 5321098 | ||
BC034059.1 | - | 10 | 1207 | 4163877 | ||
BC096743.1 | - | 16 | 2226 | 30549276 |
