siRNA Details
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007509.3 | NP_031535.2 | 7, 8 | 761 | ||
GenBank mRNA | AK146499.1 | - | 7, 8 | 726 | ||
AK151200.1 | - | 7, 8 | 762 | |||
AK151322.1 | - | 7, 8 | 734 | |||
AK151586.1 | - | 7, 8 | 734 | |||
AK152718.1 | - | 7, 8 | 734 | |||
AK152766.1 | - | 7, 8 | 734 | |||
AK153341.1 | - | 7, 8 | 725 | |||
AK159133.1 | - | 7, 8 | 732 | |||
AK159153.1 | - | 7, 8 | 732 | |||
AK159586.1 | - | 7, 8 | 729 | |||
AK159701.1 | - | 7, 8 | 733 | |||
AK159986.1 | - | 7, 8 | 733 | |||
AK160080.1 | - | 7, 8 | 732 | |||
AK160854.1 | - | 7, 8 | 727 | |||
AK166669.1 | - | 7, 8 | 701 | |||
AK168852.1 | - | 7, 8 | 725 | |||
AK169155.1 | - | 7, 8 | 740 | |||
AK169270.1 | - | 7, 8 | 737 | |||
AK169420.1 | - | 7, 8 | 729 | |||
AK170110.1 | - | 7, 8 | 756 | |||
AK196120.1 | - | 2, 3 | 180 | |||
AK207339.1 | - | 2, 3 | 180 | |||
AK207413.1 | - | 2, 3 | 180 | |||
AK209086.1 | - | 2, 3 | 180 | |||
AK210829.1 | - | 2, 3 | 180 | |||
AK210876.1 | - | 4, 5 | 376 | |||
AK215006.1 | - | 2, 3 | 180 | |||
AK216515.1 | - | 4, 5 | 369 | |||
BC012497.1 | - | 7, 8 | 726 | 4500843 | ||
BC046302.1 | - | 7, 8 | 716 | 6492612 | ||
BC085300.1 | - | 7, 8 | 708 | 6305977 | ||
Y12634.1 | - | 7, 8 | 718 |
