siRNA Details
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007509.3 | NP_031535.2 | 10 | 1036 | ||
GenBank mRNA | AK146499.1 | - | 10 | 1001 | ||
AK151200.1 | - | 10 | 1037 | |||
AK151322.1 | - | 10 | 1009 | |||
AK151586.1 | - | 10 | 1009 | |||
AK152718.1 | - | 10 | 1009 | |||
AK152766.1 | - | 10 | 1009 | |||
AK153341.1 | - | 10 | 1000 | |||
AK159133.1 | - | 10 | 1007 | |||
AK159153.1 | - | 10 | 1007 | |||
AK159586.1 | - | 10 | 1004 | |||
AK159701.1 | - | 10 | 1008 | |||
AK159986.1 | - | 10 | 1008 | |||
AK160080.1 | - | 10 | 1007 | |||
AK160854.1 | - | 10 | 1002 | |||
AK166669.1 | - | 10 | 976 | |||
AK168852.1 | - | 10 | 1000 | |||
AK169155.1 | - | 10 | 1015 | |||
AK169270.1 | - | 10 | 1012 | |||
AK169420.1 | - | 10 | 1004 | |||
AK170110.1 | - | 10 | 1031 | |||
AK198089.1 | - | 2 | 104 | |||
AK201716.1 | - | 2 | 104 | |||
AK203319.1 | - | 2 | 104 | |||
AK204547.1 | - | 2 | 104 | |||
BC012497.1 | - | 10 | 1001 | 4500843 | ||
BC046302.1 | - | 10 | 991 | 6492612 | ||
BC085300.1 | - | 10 | 983 | 6305977 | ||
Y12634.1 | - | 10 | 993 |
