siRNA Details
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013477.3 | NP_038505.2 | 4, 5 | 700 | ||
GenBank mRNA | AB088408.1 | - | 4, 5 | 589 | ||
AK083372.1 | - | 4, 5 | 701 | |||
AK133921.1 | - | 4, 5 | 708 | |||
AK171515.1 | - | 4, 5 | 670 | |||
AK213362.1 | - | 1, 2 | 91 | |||
BC011075.1 | - | 4, 5 | 591 | 3662404 | ||
U13840.1 | - | 4, 5 | 589 | |||
U21549.1 | - | 4, 5 | 572 |
