siRNA Details
Gene Symbol: Atp8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038999.2 | NP_001034088.1 | 36, 37 | 3616 | ||
NM_001284345.1 | NP_001271274.1 | 36, 37 | 3616 | |||
NM_009727.3 | NP_033857.1 | 35, 36 | 3571 | |||
GenBank mRNA | AK045367.1 | - | 36, 37 | 3597 | ||
AK076388.1 | - | 35, 36 | 3507 | |||
AK079430.1 | - | 14, 15 | 1414 | |||
AK141559.1 | - | 36, 37 | 3551 | |||
AK162598.1 | - | 4, 5 | 312 | |||
AK220560.1 | - | 36, 37 | 3526 | |||
BC094235.1 | - | 25, 26 | 2321 | |||
BC129872.1 | - | 36, 37 | 3391 | 40110214 | ||
U75321.1 | - | 35, 36 | 3350 |
