siRNA Details
Gene Symbol: Pcdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142735.1 | NP_001136207.1 | 30 | 4599 | ||
NM_001142736.1 | NP_001136208.1 | 30 | 4599 | |||
NM_001142737.1 | NP_001136209.1 | 28 | 4386 | |||
NM_001142738.1 | NP_001136210.1 | 29 | 4488 | |||
NM_001142739.1 | NP_001136211.1 | 29 | 4533 | |||
NM_001142740.1 | NP_001136212.1 | 30 | 4599 | |||
NM_001142741.1 | NP_001136213.1 | 29 | 4533 | |||
NM_001142742.1 | NP_001136214.1 | 32 | 4635 | |||
NM_001142743.1 | NP_001136215.1 | 30 | 4599 | |||
NM_001142746.1 | NP_001136218.1 | 31 | 4614 | |||
NM_001142760.1 | NP_001136232.1 | 30 | 4599 | |||
NM_023115.3 | NP_075604.2 | 31 | 4614 | |||
GenBank mRNA | AF281899.1 | - | 31 | 4598 | ||
AY949849.1 | - | 18 | 2888 | |||
DQ354396.1 | - | 30 | 4583 | |||
DQ354397.1 | - | 29 | 4503 | |||
DQ354398.1 | - | 30 | 4583 | |||
DQ354399.1 | - | 29 | 4503 | |||
DQ354400.1 | - | 28 | 4370 | |||
DQ354401.1 | - | 29 | 4472 | |||
DQ354402.1 | - | 29 | 4517 | |||
DQ354403.1 | - | 30 | 4583 | |||
DQ354404.1 | - | 29 | 4517 | |||
DQ354405.1 | - | 32 | 4619 | |||
DQ354406.1 | - | 30 | 4583 | |||
DQ354407.1 | - | 23 | 3597 | |||
DQ354408.1 | - | 10 | 1653 | |||
DQ354413.1 | - | 31 | 4598 | |||
DQ354414.1 | - | 30 | 4583 | |||
DQ354415.1 | - | 4 | 838 | |||
HQ404375.1 | - | 1 | 92 | |||
HQ420254.1 | - | 1 | 92 |
