siRNA Details
Gene Symbol: Pcdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142735.1 | NP_001136207.1 | 27 | 3974 | ||
NM_001142736.1 | NP_001136208.1 | 27 | 3974 | |||
NM_001142737.1 | NP_001136209.1 | 25 | 3761 | |||
NM_001142738.1 | NP_001136210.1 | 26 | 3863 | |||
NM_001142739.1 | NP_001136211.1 | 26 | 3908 | |||
NM_001142740.1 | NP_001136212.1 | 27 | 3974 | |||
NM_001142741.1 | NP_001136213.1 | 26 | 3908 | |||
NM_001142742.1 | NP_001136214.1 | 29 | 4010 | |||
NM_001142743.1 | NP_001136215.1 | 27 | 3974 | |||
NM_001142746.1 | NP_001136218.1 | 28 | 3989 | |||
NM_001142760.1 | NP_001136232.1 | 27 | 3974 | |||
NM_023115.3 | NP_075604.2 | 28 | 3989 | |||
GenBank mRNA | AF281899.1 | - | 28 | 3973 | ||
AY949849.1 | - | 15 | 2263 | |||
DQ354396.1 | - | 27 | 3958 | |||
DQ354397.1 | - | 26 | 3878 | |||
DQ354398.1 | - | 27 | 3958 | |||
DQ354399.1 | - | 26 | 3878 | |||
DQ354400.1 | - | 25 | 3745 | |||
DQ354401.1 | - | 26 | 3847 | |||
DQ354402.1 | - | 26 | 3892 | |||
DQ354403.1 | - | 27 | 3958 | |||
DQ354404.1 | - | 26 | 3892 | |||
DQ354405.1 | - | 29 | 3994 | |||
DQ354406.1 | - | 27 | 3958 | |||
DQ354407.1 | - | 20 | 2972 | |||
DQ354413.1 | - | 28 | 3973 | |||
DQ354414.1 | - | 27 | 3958 |
