siRNA Details
Gene Symbol: Pcdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142735.1 | NP_001136207.1 | 34 | 4920 | ||
NM_001142736.1 | NP_001136208.1 | 33 | 4914 | |||
NM_001142737.1 | NP_001136209.1 | 32 | 4707 | |||
NM_001142738.1 | NP_001136210.1 | 31 | 4794 | |||
NM_001142739.1 | NP_001136211.1 | 33 | 4854 | |||
NM_001142740.1 | NP_001136212.1 | 32 | 4905 | |||
NM_001142741.1 | NP_001136213.1 | 32 | 4845 | |||
NM_023115.3 | NP_075604.2 | 35 | 4935 | |||
GenBank mRNA | AF281899.1 | - | 35 | 4919 | ||
AY949849.1 | - | 22 | 3179 | |||
DQ354396.1 | - | 34 | 4904 | |||
DQ354397.1 | - | 33 | 4824 | |||
DQ354398.1 | - | 33 | 4898 | |||
DQ354399.1 | - | 32 | 4818 | |||
DQ354400.1 | - | 32 | 4691 | |||
DQ354401.1 | - | 31 | 4778 | |||
DQ354402.1 | - | 33 | 4838 | |||
DQ354403.1 | - | 32 | 4889 | |||
DQ354404.1 | - | 32 | 4829 |
