siRNA Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009758.4 | NP_033888.2 | 7 | 884 | ||
GenBank mRNA | AK132051.1 | - | 7 | 897 | ||
AY365062.1 | - | 5 | 478 | |||
BC042611.1 | - | 6 | 674 | 5364272 | ||
D16250.1 | - | 5 | 619 | |||
U04672.1 | - | 5 | 488 | |||
Z23154.1 | - | 5 | 478 |
