siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009760.4 | NP_033890.1 | 2 | 197 | ||
GenBank mRNA | AF041054.1 | - | 2 | 167 | ||
AK014223.1 | - | 2 | 198 | |||
AK075943.1 | - | 2 | 198 | |||
AK152610.1 | - | 2 | 193 | |||
AK170865.1 | - | 2 | 162 | |||
BC033278.1 | - | 2 | 105 | |||
BC046603.1 | - | 2 | 187 | 6390074 |
