siRNA Details
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 46, 47 | 9433 | ||
GenBank mRNA | AK052612.1 | - | 1 | 19 | ||
CN722212.1 | - | 1, 2 | 116 | |||
EL608144.1 | - | 3 | 346 | |||
Y17267.1 | - | 46, 47 | 9434 |
