siRNA Details
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 48 | 9685 | ||
GenBank mRNA | AK052612.1 | - | 2 | 271 | ||
CN722212.1 | - | 3 | 368 | |||
Y17267.1 | - | 48 | 9686 |
