siRNA Details
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 54 | 11322 | ||
GenBank mRNA | AK052612.1 | - | 8 | 1908 | ||
AK083695.1 | - | 1 | 397 | |||
Y17267.1 | - | 54 | 11323 |
