siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159533.2 | NP_001153005.1 | 29 | 4278 | ||
NM_001159534.2 | NP_001153006.1 | 29 | 4278 | |||
NM_001159535.2 | NP_001153007.1 | 29 | 4278 | |||
NM_001255997.2 | NP_001242926.1 | 29 | 4026 | |||
NM_001255998.2 | NP_001242927.1 | 30 | 4101 | |||
NM_001255999.2 | NP_001242928.1 | 29 | 4026 | |||
NM_001256000.2 | NP_001242929.1 | 30 | 4353 | |||
NM_001256001.2 | NP_001242930.1 | 30 | 4362 | |||
NM_001256002.2 | NP_001242931.1 | 30 | 4353 | |||
NM_001290335.1 | NP_001277264.1 | 25 | 3375 | |||
NM_009781.4 | NP_033911.2 | 29 | 4026 | |||
GenBank mRNA | AB259049.1 | - | 29 | 3848 | ||
AY728090.1 | - | 29 | 3813 | |||
BC138031.1 | - | 29 | 3972 | 8861050 | ||
BC145103.1 | - | 29 | 3972 | |||
BC145104.1 | - | 29 | 3972 | 9053652 | ||
BC145105.1 | - | 29 | 3972 | 9053653 | ||
FM872409.1 | - | 29 | 3848 | |||
FM872410.1 | - | 30 | 3923 | |||
FM872411.1 | - | 29 | 3848 | |||
FM872412.1 | - | 30 | 3833 | |||
FM872413.1 | - | 30 | 3842 | |||
FM872414.1 | - | 30 | 3833 | |||
M57973.1 | - | 3 | 275 | |||
U17869.1 | - | 25 | 3375 |
