siRNA Details
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 9 | 1269 | ||
NM_001291234.1 | NP_001278163.1 | 9 | 980 | |||
NM_001291238.1 | NP_001278167.1 | 9 | 980 | |||
NM_001291239.1 | NP_001278168.1 | 9 | 1269 | |||
NM_001291240.1 | NP_001278169.1 | 9 | 1269 | |||
NM_009986.4 | NP_034116.3 | 9 | 980 | |||
NM_198602.3 | NP_941004.2 | 9 | 980 | |||
GenBank mRNA | AB041595.1 | - | 9 | 1119 | ||
AF004225.1 | - | 5 | 339 | |||
AK098096.1 | - | 9 | 743 | |||
AK144932.1 | - | 9 | 1640 | |||
AK220173.1 | - | 9 | 736 | |||
AY037807.1 | - | 9 | 710 | |||
BC014289.1 | - | 9 | 749 | |||
BC054370.1 | - | 9 | 703 | |||
BC079570.1 | - | 10 | 1167 | |||
BC090847.1 | - | 9 | 752 | 30547458 | ||
U68542.1 | - | 10 | 754 | |||
X75013.1 | - | 5 | 470 |
