siRNA Details
Gene Symbol: Cyp7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007824.2 | NP_031850.2 | 1 | 101 | ||
GenBank mRNA | AK050020.1 | - | 1 | 101 | ||
AK050210.1 | - | 1 | 102 | |||
AK050220.1 | - | 1 | 101 | |||
AK050260.1 | - | 1 | 101 | |||
BC130261.1 | - | 1 | 71 | 40142789 |
