siRNA Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008702.2 | NP_001008702.1 | 11 | 1198 | ||
NM_001037905.3 | NP_001032994.1 | 11 | 1272 | |||
NM_001102400.1 | NP_001095870.1 | 10 | 1209 | |||
NM_023118.5 | NP_075607.2 | 12 | 1926 | |||
GenBank mRNA | AK154716.1 | - | 12 | 1910 | ||
AK162744.1 | - | 10 | 1180 | |||
AK162851.1 | - | 11 | 1243 | |||
AK165519.1 | - | 11 | 1156 | |||
AK165810.1 | - | 11 | 1153 | |||
AK168375.1 | - | 11 | 1245 | |||
BC016887.1 | - | 11 | 1138 | 4242174 | ||
U18869.1 | - | 12 | 1859 |
