siRNA Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 2 | 142 | ||
GenBank mRNA | AB018002.1 | - | 2 | 132 | ||
AK004350.1 | - | 2 | 115 | |||
AK158082.1 | - | 2 | 134 | |||
BC022165.1 | - | 2 | 111 | 4016911 | ||
CK032192.1 | - | 2 | 175 | |||
GU109477.1 | - | 2 | 142 |
