siRNA Details
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007885.2 | NP_031911.1 | 3 | 921 | ||
GenBank mRNA | AK007212.1 | - | 3 | 987 | ||
AK052942.1 | - | 3 | 922 | |||
AK163974.1 | - | 3 | 812 | |||
BC028345.1 | - | 3 | 798 | 4014785 | ||
D42049.1 | - | 3 | 806 |
