siRNA Details
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291105.1 | NP_001278034.1 | 6, 7 | 1159 | ||
NM_007891.5 | NP_031917.1 | 6, 7 | 1203 | |||
GenBank mRNA | AF483514.1 | - | 6, 7 | 1045 | ||
AF483515.1 | - | 6, 7 | 1045 | |||
AK017841.1 | - | 6, 7 | 1151 | |||
AK153968.1 | - | 6, 7 | 1168 | |||
AK220226.1 | - | 6, 7 | 900 | |||
BC052160.1 | - | 6, 7 | 1145 | 6330425 | ||
L21973.1 | - | 6, 7 | 1163 |
