siRNA Details
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291105.1 | NP_001278034.1 | 5 | 920 | ||
NM_007891.5 | NP_031917.1 | 5 | 964 | |||
GenBank mRNA | AF483514.1 | - | 5 | 806 | ||
AF483515.1 | - | 5 | 806 | |||
AK017841.1 | - | 5 | 912 | |||
AK153968.1 | - | 5 | 929 | |||
AK182005.1 | - | 2 | 218 | |||
AK211069.1 | - | 3 | 317 | |||
AK220226.1 | - | 5 | 661 | |||
BC052160.1 | - | 5 | 906 | 6330425 | ||
L21973.1 | - | 5 | 924 |
