siRNA Details
Gene Symbol: Ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276387.1 | NP_001263316.1 | 13 | 2120 | ||
NM_010095.6 | NP_034225.1 | 13 | 2115 | |||
GenBank mRNA | AK053013.1 | - | 13 | 2116 | ||
AK139262.1 | - | 13 | 1827 | |||
BC049188.1 | - | 12 | 1414 | 6312260 | ||
BC050922.1 | - | 12 | 1637 | 6409737 | ||
U71189.1 | - | 12 | 1300 | |||
U82441.1 | - | 12 | 1169 | |||
U92703.1 | - | 12 | 1520 |
