siRNA Details
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136061.2 | NP_001129533.1 | 2 | 678 | ||
NM_001276296.1 | NP_001263225.1 | 1 | 492 | |||
NM_007904.4 | NP_031930.1 | 1 | 492 | |||
GenBank mRNA | AK076426.1 | - | 1 | 486 | ||
AK082103.1 | - | 2 | 479 | |||
AK083415.1 | - | 2 | 375 | |||
AK085165.1 | - | 1 | 483 | |||
AK085532.1 | - | 1 | 488 | |||
BC026553.1 | - | 2 | 604 | 4971909 | ||
BQ442271.1 | - | 1 | 493 | |||
U32329.1 | - | 1 | 470 |
