siRNA Details
Gene Symbol: Efna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010109.3 | NP_034239.1 | 4 | 763 | ||
NM_207654.2 | NP_997537.1 | 5 | 844 | |||
GenBank mRNA | BC040218.1 | - | 5 | 844 | 4954352 | |
U90664.1 | - | 5 | 671 | |||
U90665.1 | - | 4 | 590 |
