siRNA Details
Gene Symbol: Etv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035231.1 | - | 7 | 1288 | ||
AK078773.1 | - | 8 | 1481 | |||
AK154569.1 | - | 8 | 1569 | |||
AK155150.1 | - | 8 | 1754 | |||
AK155752.1 | - | 7 | 1304 | |||
AK157265.1 | - | 8 | 1423 | |||
BC052163.1 | - | 8 | 1442 | 6513848 |
