siRNA Details
Gene Symbol: Strc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080459.2 | NP_536707.2 | 27, 28 | 5267 | ||
GenBank mRNA | AF375593.1 | - | 27, 28 | 5267 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Strc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080459.2 | NP_536707.2 | 27, 28 | 5267 | ||
GenBank mRNA | AF375593.1 | - | 27, 28 | 5267 |