siRNA Details
Gene Symbol: Man2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172903.4 | NP_766491.2 | 3, 4 | 634 | ||
GenBank mRNA | AK029913.1 | - | 3, 4 | 445 | ||
AK044028.1 | - | 2, 3 | 1372 | |||
AK161799.1 | - | 2, 3 | 293 | |||
DQ631806.1 | - | 1, 2 | 149 |
