siRNA Details
Gene Symbol: Atp6v0a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080467.3 | NP_536715.3 | 11 | 1377 | ||
GenBank mRNA | AB050903.1 | - | 10 | 1103 | ||
AF326316.1 | - | 11 | 1171 | |||
AF435090.1 | - | 11 | 1365 | |||
AK128892.1 | - | 11 | 1377 | |||
BC046979.1 | - | 12 | 1399 | 6315485 |
