siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004817.1 | - | 16 | 1915 | ||
AF483490.1 | - | 14 | 1804 | |||
AF483491.1 | - | 14 | 1804 | |||
AK129004.1 | - | 15 | 1875 | |||
AK138942.1 | - | 16 | 1898 | |||
AK140694.1 | - | 16 | 1922 | |||
AK154565.1 | - | 16 | 1895 | |||
AK211176.1 | - | 3 | 311 | |||
AK212970.1 | - | 2 | 287 | |||
BC007135.1 | - | 15 | 1875 | 3600884 | ||
U60453.1 | - | 16 | 1903 |

More Information
Other Species Targeted

