siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004817.1 | - | 8 | 846 | ||
AF483490.1 | - | 6 | 735 | |||
AF483491.1 | - | 6 | 735 | |||
AK045374.1 | - | 8 | 834 | |||
AK129004.1 | - | 7 | 806 | |||
AK138942.1 | - | 8 | 829 | |||
AK140694.1 | - | 8 | 853 | |||
AK154565.1 | - | 8 | 826 | |||
AK164192.1 | - | 8 | 845 | |||
BC007135.1 | - | 7 | 806 | 3600884 | ||
U60453.1 | - | 8 | 834 |
