siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004817.1 | - | 17 | 2007 | ||
AF483490.1 | - | 15 | 1896 | |||
AF483491.1 | - | 15 | 1896 | |||
AK129004.1 | - | 16 | 1967 | |||
AK138942.1 | - | 17 | 1990 | |||
AK140694.1 | - | 17 | 2014 | |||
AK154565.1 | - | 17 | 1987 | |||
AK185194.1 | - | 1 | 19 | |||
BC007135.1 | - | 16 | 1967 | 3600884 | ||
U60453.1 | - | 17 | 1995 |

More Information
Other Species Targeted

