siRNA Details
Gene Symbol: Fbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019395.3 | NP_062268.1 | 1, 2 | 414 | ||
GenBank mRNA | AJ132693.1 | - | 1, 2 | 365 | ||
AK002216.1 | - | 1, 2 | 276 | |||
AK149527.1 | - | 1, 2 | 295 | |||
BC011480.1 | - | 1, 2 | 278 | 4194764 | ||
BC051392.1 | - | 1, 2 | 278 | 5054854 |
