siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079908.2 | NP_001073377.1 | 15 | 2771 | ||
NM_001079909.2 | NP_001073378.1 | 14 | 2510 | |||
NM_010206.3 | NP_034336.2 | 15 | 2777 | |||
GenBank mRNA | AF176552.1 | - | 13 | 1808 | ||
AK028354.1 | - | 14 | 1950 | |||
AK034295.1 | - | 5 | 597 | |||
AK049704.1 | - | 15 | 2737 | |||
AK167639.1 | - | 6 | 2074 | |||
BC010200.1 | - | 15 | 2670 | 3967874 | ||
BC033447.1 | - | 13 | 1773 | 4037899 | ||
BC053921.1 | - | 8 | 2017 | |||
M28998.1 | - | 14 | 2075 | |||
M33760.1 | - | 13 | 1794 | |||
M65053.1 | - | 14 | 2048 | |||
U22324.1 | - | 14 | 2075 | |||
U23445.1 | - | 13 | 1808 | |||
X51893.1 | - | 15 | 2132 |
