siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079908.2 | NP_001073377.1 | 9 | 1967 | ||
NM_001079909.2 | NP_001073378.1 | 8 | 1706 | |||
NM_010206.3 | NP_034336.2 | 9 | 1973 | |||
GenBank mRNA | AF176552.1 | - | 7 | 1004 | ||
AK028354.1 | - | 8 | 1146 | |||
AK049704.1 | - | 9 | 1933 | |||
AK086091.1 | - | 1 | 843 | |||
BC010200.1 | - | 9 | 1866 | 3967874 | ||
BC033447.1 | - | 7 | 969 | 4037899 | ||
BC053921.1 | - | 2 | 1213 | |||
M28998.1 | - | 8 | 1271 | |||
M33760.1 | - | 7 | 990 | |||
M65053.1 | - | 8 | 1244 | |||
U22324.1 | - | 8 | 1271 | |||
U23445.1 | - | 7 | 1004 | |||
X51893.1 | - | 9 | 1328 |
