siRNA Details
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010220.4 | NP_034350.1 | 6 | 761 | ||
GenBank mRNA | AK169795.1 | - | 6 | 702 | ||
BC015260.1 | - | 6 | 700 | 4237766 | ||
BU613862.1 | - | 4 | 428 | |||
CA317455.1 | - | 6 | 724 | |||
CT010372.1 | - | 5 | 542 | |||
U16959.1 | - | 6 | 761 | |||
U36220.1 | - | 6 | 689 |
