siRNA Details
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080845.2 | NP_543121.1 | 10, 11 | 1312 | ||
GenBank mRNA | AK165414.1 | - | 8, 9 | 1519 | ||
BC010813.1 | - | 10, 11 | 1274 | 4211065 | ||
BC024078.1 | - | 10, 11 | 1306 | 5098529 | ||
CB949427.1 | - | 4, 5 | 450 |
