siRNA Details
Gene Symbol: Ftcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080845.2 | NP_543121.1 | 13, 14 | 1607 | ||
GenBank mRNA | AK165414.1 | - | 11, 12 | 1814 | ||
BC010813.1 | - | 13, 14 | 1569 | 4211065 | ||
BC024078.1 | - | 13, 14 | 1601 | 5098529 |
