siRNA Details
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008087.2 | NP_032113.1 | 6 | 809 | ||
GenBank mRNA | AK014509.1 | - | 8 | 1314 | ||
AK142903.1 | - | 8 | 988 | |||
AK188527.1 | - | 3 | 318 | |||
BC013456.1 | - | 5 | 577 | 4237356 | ||
BC053446.1 | - | 6 | 858 | 6336752 | ||
M21828.1 | - | 6 | 809 |
