siRNA Details
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008087.2 | NP_032113.1 | 7, 8 | 969 | ||
GenBank mRNA | AK014509.1 | - | 9, 10 | 1474 | ||
BC013456.1 | - | 6, 7 | 737 | 4237356 | ||
BC053446.1 | - | 7, 8 | 1018 | 6336752 | ||
M21828.1 | - | 7, 8 | 969 |
