siRNA Details
Gene Symbol: Gcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008103.3 | NP_032129.2 | 2 | 201 | ||
GenBank mRNA | AK142574.1 | - | 2 | 187 | ||
BC066866.1 | - | 2 | 201 | 30432193 | ||
D88612.1 | - | 2 | 236 | |||
U59876.1 | - | 2 | 217 |
