siRNA Details
Gene Symbol: Ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010281.2 | NP_034411.2 | 5 | 598 | ||
GenBank mRNA | AF051102.1 | - | 5 | 566 | ||
AI846732.1 | - | 2 | 154 | |||
AK089996.1 | - | 5 | 555 | |||
BB610405.1 | - | 5 | 486 | |||
BC009809.1 | - | 5 | 489 | 4223953 | ||
BC150898.1 | - | 5 | 535 | 9087809 |
