siRNA Details
Gene Symbol: Gh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008117.3 | NP_032143.1 | 3 | 349 | ||
GenBank mRNA | AK019952.1 | - | 3 | 264 | ||
AK019954.1 | - | 3 | 264 | |||
AK019956.1 | - | 3 | 261 | |||
AK019959.1 | - | 3 | 264 | |||
AK019961.1 | - | 3 | 264 | |||
AK030419.1 | - | 3 | 316 | |||
AK030715.1 | - | 3 | 264 | |||
BC061157.1 | - | 3 | 263 | 30250975 | ||
X02891.1 | - | 3 | 263 |
