siRNA Details
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281969.1 | NP_001268898.1 | 5 | 501 | ||
NM_010298.6 | NP_034428.2 | 5 | 551 | |||
GenBank mRNA | AK082578.1 | - | 5 | 553 | ||
AK083251.1 | - | 4 | 405 | |||
AM420316.1 | - | 4 | 384 | |||
BC027094.1 | - | 5 | 505 | |||
BC037605.1 | - | 5 | 519 | 5361798 | ||
U09399.1 | - | 5 | 460 | |||
X81202.1 | - | 4 | 343 | |||
X81203.1 | - | 2 | 126 |
