siRNA Details
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281969.1 | NP_001268898.1 | 9, 10 | 1327 | ||
NM_010298.6 | NP_034428.2 | 9, 10 | 1377 | |||
GenBank mRNA | AK082578.1 | - | 9, 10 | 1380 | ||
AK083251.1 | - | 8, 9 | 1231 | |||
AM420316.1 | - | 7, 8 | 1057 | |||
BC027094.1 | - | 9, 10 | 1331 | |||
BC037605.1 | - | 9, 10 | 1345 | 5361798 | ||
U09399.1 | - | 9, 10 | 1286 | |||
X81202.1 | - | 8, 9 | 1169 |
