siRNA Details
Gene Symbol: Pdpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290822.1 | NP_001277751.1 | 4 | 707 | ||
NM_010329.3 | NP_034459.2 | 5 | 746 | |||
GenBank mRNA | AJ250246.1 | - | 5 | 475 | ||
AJ297944.2 | - | 5 | 452 | |||
AK158855.1 | - | 5 | 746 | |||
BC026551.1 | - | 5 | 630 | 4985288 | ||
CX240531.1 | - | 4 | 606 | |||
M73748.1 | - | 5 | 603 | |||
M96645.1 | - | 5 | 513 |
