siRNA Details
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010431.2 | NP_034561.2 | 7 | 1246 | ||
GenBank mRNA | AF003695.1 | - | 7 | 1080 | ||
AK034087.1 | - | 7 | 1212 | |||
AK076395.1 | - | 5 | 534 | |||
AK138944.1 | - | 7 | 1113 | |||
AK150367.1 | - | 7 | 1113 | |||
BC026139.1 | - | 7 | 1098 | 4019056 | ||
X95580.1 | - | 7 | 912 |
