siRNA Details
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010431.2 | NP_034561.2 | 15 | 2916 | ||
GenBank mRNA | AF003695.1 | - | 15 | 2750 | ||
AK033471.1 | - | 4 | 516 | |||
AK034087.1 | - | 15 | 2882 | |||
AK076395.1 | - | 13 | 2204 | |||
AK150367.1 | - | 15 | 2783 | |||
BC026139.1 | - | 15 | 2768 | 4019056 | ||
X95580.1 | - | 15 | 2540 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_024359, XM_006240197, XM_006240196, XM_006240199, XM_008764695, and others.
Gene Ontology Categories:
